4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzofuran 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Dibenzo[b,d]furan-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, Dibenzofuran-4-boronic acid Pinacol ester, 2-(Dibenzofuran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A382449-100MG A382449-1G A382449-250MG
76990-006EA 8 USD
76990-006 76990-008 76990-010
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzofuran 97%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)dibenzofuran

Precaución: for Research Use Only

Formula: C₁₈H₁₉BO₃
MDL Number: MFCD18910632
CAS Number: 912824-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR