1-Ethyl-3-nitrobenzene 95%
Supplier: Ambeed
|
Warning
|
A382128-100MG
A382128-1G
A382128-250MG
77016-074EA
130.5
USD
77016-074
77016-076
77684-101
1-Ethyl-3-nitrobenzene 95%
1-Ethyl-3-nitrobenzene
Precaución: For research use only.
Formula:
C₈H₉NO₂ MW: 151.16 g/mol |
MDL Number:
MFCD00130058 CAS Number: 7369-50-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...