2-(6-Chloropyridin-2-yl)acetic acid 95%

Supplier: Ambeed
Warning

A381930-100MG A381930-250MG A381930-5G A381930-1G
77033-432EA 44.56 USD
77033-432 77033-434 77033-436 77668-778
2-(6-Chloropyridin-2-yl)acetic acid 95%
2-(6-Chloropyridin-2-yl)acetic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
MDL Number: MFCD08706288
CAS Number: 885267-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR