3-Bromo-4-chlorobenzeneacetic acid 97%

Supplier: Ambeed
Warning

A381824-100G A381824-10G A381824-1G A381824-250MG A381824-5G A381824-25G
76989-996EA 210 USD
76989-996 76989-998 76990-000 76990-002 76990-004 77886-003
3-Bromo-4-chlorobenzeneacetic acid 97%
3-Bromo-4-chlorobenzeneacetic acid

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
MDL Number: MFCD18390582
CAS Number: 90004-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR