4-Morpholin-4-yl-2-nitrobenzoic acid 95%

Supplier: Ambeed
Warning

A381446-100MG A381446-10G A381446-1G A381446-250MG A381446-5G
77033-420EA 37 USD
77033-420 77033-422 77033-424 77033-426 77033-428
4-Morpholin-4-yl-2-nitrobenzoic acid 95%
4-Morpholin-4-yl-2-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₅
MDL Number: MFCD12198419
CAS Number: 404010-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR