tert-Butyl 3-cyano-3-hydroxypyrrolidine-1-carboxylate 97%

Supplier: Ambeed
Warning

A381423-1G A381423-250MG
77033-416EA 329.94 USD
77033-416 77033-418
tert-Butyl 3-cyano-3-hydroxypyrrolidine-1-carboxylate 97%
tert-Butyl 3-cyano-3-hydroxypyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₃
MW: 212.25 g/mol
MDL Number: MFCD11878020
CAS Number: 1194376-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR