4-(Dimethylamino)butan-2-one 95%

Supplier: Ambeed
Danger

A380968-5G A380968-1G A380968-25G
77003-640EA 189 USD
77003-640 77568-766 77882-019
4-(Dimethylamino)butan-2-one 95%
4-(Dimethylamino)butan-2-one

Precaución: For research use only.

Formula: C₆H₁₃NO
MDL Number: MFCD00457365
CAS Number: 2543-57-9
UN: 2733
ADR: 3,8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR