N-Boc-4-nitroaniline 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (4-nitrophenyl)carbamate

A380928-100G A380928-25G A380928-5G
77016-066EA 387.23 USD
77016-066 77016-068 77016-070
N-Boc-4-nitroaniline 97%
N-Boc-4-nitroaniline

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O₄
MW: 238.2399 g/mol
MDL Number: MFCD01006654
CAS Number: 18437-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR