FK228 98%

Supplier: Ambeed
Warning

Synonyms: Romidepsin, (1S,4S,7Z,10S,16E,21R)-7-Ethylidene-4,21-diisopropyl-2-oxa-12,13-dithia-5,8,20,23-tetraazabicyclo[8.7.6]tricos-16-ene-3,6,9,19,22-pentaone

A380298-10MG A380298-1MG A380298-5MG
76989-974EA 593.05 USD
76989-974 76989-976 76989-978
FK228 98%
FK228

Precaución: for Research Use Only

Formula: C₂₄H₃₆N₄O₆S₂
MW: 540.7 g/mol
MDL Number: MFCD18433404
CAS Number: 128517-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR