4-Chloro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine 98%

Supplier: Ambeed
Warning

A380257-100MG A380257-1G A380257-250MG A380257-5G
77033-408EA 13.8 USD
77033-408 77033-410 77033-412 77033-414
4-Chloro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine 98%
4-Chloro-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₁₆H₂₅ClN₂Si
MDL Number: MFCD09763673
CAS Number: 651744-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR