3-Methoxy-4-(trifluoromethyl)phenylamine 98%

Supplier: Ambeed
Warning

A379777-250MG A379777-5G A379777-1G
77003-636EA 35 USD
77003-636 77003-638 77601-672
3-Methoxy-4-(trifluoromethyl)phenylamine 98%
3-Methoxy-4-(trifluoromethyl)phenylamine

Precaución: for Research Use Only

Formula: C₈H₈F₃NO
MDL Number: MFCD11840978
CAS Number: 106877-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR