2-Bromo-1,3-cyclopentanedione 98%

Supplier: Ambeed
Warning

A379764-100MG A379764-1G A379764-250MG
77003-630EA 18 USD
77003-630 77003-632 77003-634
2-Bromo-1,3-cyclopentanedione 98%
2-Bromo-1,3-cyclopentanedione

Precaución: for Research Use Only

Formula: C₅H₅BrO₂
MDL Number: MFCD12827833
CAS Number: 14203-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR