1-N-BOC-3-methyl-indazole-5-boronic acid, pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate

A379570-100MG A379570-1G A379570-250MG
77033-400EA 45 USD
77033-400 77033-402 77033-404
1-N-BOC-3-methyl-indazole-5-boronic acid, pinacol ester 97%
1-N-BOC-3-methyl-indazole-5-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₇BN₂O₄
MDL Number: MFCD13195073
CAS Number: 864770-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR