9-Ethynylphenanthrene 95%

Supplier: Ambeed
Danger

A379320-250MG A379320-5G A379320-1G A379320-25G
77033-398EA 11 USD
77033-398 77601-666 77668-756 77872-589
9-Ethynylphenanthrene 95%
9-Ethynylphenanthrene

Precaución: for Research Use Only

Formula: C₁₆H₁₀
MDL Number: MFCD05664210
CAS Number: 32870-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR