2,3-Dichloro-5-(chloromethyl)pyridine 95%

Supplier: Ambeed
Danger

Synonyms: 5-(Chloromethyl)-2,3-dichloropyridine

A378973-1G A378973-250MG
76989-966EA 194 USD
76989-966 76989-968
2,3-Dichloro-5-(chloromethyl)pyridine 95%
2,3-Dichloro-5-(chloromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₄Cl₃N
MDL Number: MFCD00125365
CAS Number: 54127-31-0
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR