3-(3-Fluoro-5-nitrophenoxy)pyridine 95%

Supplier: Ambeed
Warning

A378940-1G A378940-250MG
76989-962EA 49.87 USD
76989-962 76989-964
3-(3-Fluoro-5-nitrophenoxy)pyridine 95%
3-(3-Fluoro-5-nitrophenoxy)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₇FN₂O₃
MW: 234.18 g/mol
MDL Number: MFCD18262282
CAS Number: 803700-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR