Methyl 6-bromo-2-methoxynicotinate 98%

Supplier: Ambeed
Warning

A378813-100MG A378813-10G A378813-1G A378813-250MG A378813-5G
77016-046EA 45 USD
77016-046 77016-048 77016-050 77016-052 77016-054
Methyl 6-bromo-2-methoxynicotinate 98%
Methyl 6-bromo-2-methoxynicotinate

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MDL Number: MFCD18257518
CAS Number: 1009735-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR