SCH772984 98%

Supplier: Ambeed
Warning

Synonyms: SCH-772984, (R)-1-(2-Oxo-2-(4-(4-(pyrimidin-2-yl)phenyl)piperazin-1-yl)ethyl)-N-(3-(pyridin-4-yl)-1H-indazol-5-yl)pyrrolidine-3-carboxamide, (3R)-1-[2-oxo-2-[4-(4-pyrimidin-2-ylphenyl)piperazin-1-yl]ethyl]-N-(3-pyridin-4-yl-1H-indazol-5-yl)pyrrolidine-3-carboxamide, SCH 772984, SCH772984

A378792-10MG A378792-1MG A378792-2MG A378792-50MG A378792-5MG
76989-952EA 170.81 USD
76989-952 76989-954 76989-956 76989-958 76989-960
SCH772984 98%
SCH772984

Precaución: for Research Use Only

Formula: C₃₃H₃₃N₉O₂
MW: 587.67 g/mol
MDL Number: MFCD28144700
CAS Number: 942183-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR