4-(1H-Imidazol-1-yl)benzonitrile 98%

Supplier: Ambeed
Warning

A378206-10G A378206-1G A378206-250MG A378206-25G A378206-5G
77003-618EA 103.97 USD
77003-618 77003-620 77003-622 77003-624 77003-626
4-(1H-Imidazol-1-yl)benzonitrile 98%
4-(1H-Imidazol-1-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₀H₇N₃
MW: 169.1800 g/mol
MDL Number: MFCD00127147
CAS Number: 25372-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR