5-Iodo-2-methoxypyrimidine 97%

Supplier: Ambeed
Warning

A378121-100MG A378121-1G A378121-250MG A378121-5G
77016-040EA 16.98 USD
77016-040 77016-042 77016-044 77684-094
5-Iodo-2-methoxypyrimidine 97%
5-Iodo-2-methoxypyrimidine

Precaución: For research use only.

Formula: C₅H₅IN₂O
MW: 236.01 g/mol
MDL Number: MFCD18412677
CAS Number: 101803-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR