(3-(Dimethylamino)propyl)trimethoxysilane 98% (by GC)

Supplier: Ambeed
Danger

A378115-10G A378115-1G A378115-5G A378115-25G
77033-376EA 45 USD
77033-376 77033-378 77033-380 77551-096
(3-(Dimethylamino)propyl)trimethoxysilane 98% (by GC)
(3-(Dimethylamino)propyl)trimethoxysilane

Precaución: for Research Use Only

Formula: (H₃C)₂NCH₂CH₂CH₂Si(OCH₃)₃
MDL Number: MFCD00053674
CAS Number: 2530-86-1
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR