Methyl-3-iodo-1H-indazole-6-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A378092-10G
A378092-1G
A378092-250MG
77033-370EA
91
USD
77033-370
77033-372
77033-374
Methyl-3-iodo-1H-indazole-6-carboxylate 95%
Methyl 3-iodo-1H-indazole-6-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₇IN₂O₂ |
MDL Number:
MFCD07781616 CAS Number: 885518-82-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...