8-Azaguanine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Amino-3,6-dihydro-[1,2,3]triazolo[4,5-d]pyrimidin-7-one, 2-Amino-6-hydroxy-8-azapurine, 5-Amino-1,4-dihydro-7H-[1,2,3]triazolo[4,5-d]pyrimidin-7-one, Azaguanine-8, Pathocidine, 8-Azaguanine, Guanazolo, 5-Amino-1H-triazolo[4,5-d]pyrimidin-7-ol

A378059-25MG A378059-5G A378059-1MG
77003-614EA 6 USD
77003-614 77003-616 77594-136
8-Azaguanine 95%
8-Azaguanine

Precaución: for Research Use Only

Formula: C₄H₄N₆O
MW: 152.12 g/mol
MDL Number: MFCD00056937
CAS Number: 134-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR