6-Chloropyridazine-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A377581-100MG A377581-1G A377581-250MG A377581-5G
76989-944EA 9 USD
76989-944 76989-946 76989-948 76989-950
6-Chloropyridazine-4-carboxylic acid 95%
6-Chloropyridazine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₃ClN₂O₂
MDL Number: MFCD15146792
CAS Number: 1256794-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR