(2R,4R)-4-Methylpiperidine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A377407-10G A377407-1G A377407-250MG A377407-5G
77033-362EA 377.69 USD
77033-362 77033-364 77033-366 77033-368
(2R,4R)-4-Methylpiperidine-2-carboxylic acid 98%
(2R,4R)-4-Methylpiperidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₃NO₂
MW: 143.1800 g/mol
MDL Number: MFCD03839858
CAS Number: 74892-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR