1-Methyl-1H-indazole-4-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (1-Methyl-1H-indazol-4-yl)boronic acid

A377302-5G A377302-1G
77033-360EA 144 USD
77033-360 77872-585
1-Methyl-1H-indazole-4-boronic acid 98%
1-Methyl-1H-indazole-4-boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BN₂O₂
MW: 175.9803 g/mol
MDL Number: MFCD09870045
CAS Number: 1001907-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR