7-Methoxy-6-nitroquinazolin-4(3H)-one 95%

Supplier: Ambeed
Warning

A377289-100MG A377289-1G A377289-250MG
77033-354EA 9.55 USD
77033-354 77033-356 77033-358
7-Methoxy-6-nitroquinazolin-4(3H)-one 95%
7-Methoxy-6-nitroquinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₉H₇N₃O₄
MW: 221.17 g/mol
MDL Number: MFCD11869863
CAS Number: 1012057-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR