α-Ketoglutaric acid disodium salt dihydrate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Oxoglutaric acid disodium salt dihydrate, α-Ketoglutaric acid disodium salt dihydrate, Sodium α-ketoglutarate dibasic dihydrate
A377288-100G
A377288-25G
A377288-500G
77003-600EA
74
USD
77003-600
77003-602
77003-604
α-Ketoglutaric acid disodium salt dihydrate 97%
α-Ketoglutaric acid disodium salt dihydrate
Precaución: for Research Use Only
Formula:
C₅H₄O₅·2Na·2H₂O MW: 226.092 g/mol |
MDL Number:
MFCD00150702 CAS Number: 1282616-74-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...