Deferoxamine mesylate 98%

Supplier: Ambeed
Warning

A377160-1MG A377160-250MG A377160-5G
77003-592EA 10 USD
77003-592 77882-011 77882-012
Deferoxamine mesylate 98%
Deferoxamine mesylate

Precaución: for Research Use Only

Formula: C₂₆H₅₂N₆O₁₁S
MDL Number: MFCD00058605
CAS Number: 138-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR