Bromopyruvic acid 96%

Supplier: Ambeed
Danger

Synonyms: 3-Bromo-2-oxopropionic acid, 3-Bromopyruvic acid

A376712-1MG A376712-25G A376712-5G A376712-10G
77003-582EA 5.31 USD
77003-582 77003-584 77003-586 77601-662
Bromopyruvic acid 96%
Bromopyruvic acid

Precaución: for Research Use Only

Formula: BrH₂CC(O)COOH
MW: 166.96 g/mol
MDL Number: MFCD00002587
CAS Number: 1113-59-3
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR