3,5-Dibromo-2-methoxy-4-methylpyridine 95%

Supplier: Ambeed
Warning

A376167-100MG A376167-1G A376167-250MG A376167-5G
76989-936EA 20.16 USD
76989-936 76989-938 76989-940 76989-942
3,5-Dibromo-2-methoxy-4-methylpyridine 95%
3,5-Dibromo-2-methoxy-4-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₇Br₂NO
MW: 280.95 g/mol
MDL Number: MFCD26743694
CAS Number: 1332324-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR