6-Chloro-3-methyl-1H-indole 97%

Supplier: Ambeed
Warning

A376071-100MG A376071-250MG
76989-932EA 151.71 USD
76989-932 76989-934
6-Chloro-3-methyl-1H-indole 97%
6-Chloro-3-methyl-1H-indole

Precaución: for Research Use Only

Formula: C₉H₈ClN
MW: 165.61 g/mol
MDL Number: MFCD12924677
CAS Number: 158905-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR