8-Bromonaphthalen-2-amine 95%

Supplier: Ambeed
Warning

A375747-100MG A375747-250MG A375747-1G A375747-5G
76989-920EA 109 USD
76989-920 76989-922 77890-771 77890-772
8-Bromonaphthalen-2-amine 95%
8-Bromonaphthalen-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₈BrN
MDL Number: MFCD18409817
CAS Number: 167275-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR