5-(4-Hydroxyphenyl)isoxazole-3-carboxylic acid 96%

Supplier: Ambeed
Warning

A375515-100MG A375515-1G A375515-250MG A375515-5G
77003-564EA 32.89 USD
77003-564 77003-566 77003-568 77003-570
5-(4-Hydroxyphenyl)isoxazole-3-carboxylic acid 96%
5-(4-Hydroxyphenyl)isoxazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₄
MW: 205.17 g/mol
MDL Number: MFCD05237205
CAS Number: 33282-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR