6H-Indolo[2,3-b]quinoxaline 99%

Supplier: Ambeed
Warning

A375310-1G A375310-250MG
77003-560EA 64.72 USD
77003-560 77003-562
6H-Indolo[2,3-b]quinoxaline 99%
6H-Indolo[2,3-b]quinoxaline

Precaución: for Research Use Only

Formula: C₁₄H₉N₃
MDL Number: MFCD00068754
CAS Number: 84055-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR