O-tert-Butyl-L-threonine-tert-butylester acetate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Thr(tBu)-OtBu·AcOH, (2S,3R)-tert-Butyl 2-amino-3-(tert-butoxy)butanoate acetate
A375300-25G
A375300-5G
77016-024EA
77.45
USD
77016-024
77684-082
O-tert-Butyl-L-threonine-tert-butylester acetate 98%
O-tert-Butyl-L-threonine-tert-butylester acetate
Precaución: For research use only.
Formula:
C₁₄H₂₉NO₅ MW: 291.4 g/mol |
MDL Number:
MFCD00145051 CAS Number: 5854-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...