2-Acetoxybenzoic anhydride 95%

Supplier: Ambeed
Warning

Synonyms: Aspirin Impurity F

A374981-1G A374981-100MG A374981-250MG
77033-330EA 79.57 USD
77033-330 77601-658 77601-660
2-Acetoxybenzoic anhydride 95%
2-Acetoxybenzoic anhydride

Precaución: for Research Use Only

Formula: C₁₈H₁₄O₇
MW: 342.29 g/mol
MDL Number: MFCD00013236
CAS Number: 1466-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR