(4R,12aS)-N-(2,4-Difluorobenzyl)-7-hydroxy-4-methyl-6,8-dioxo-3,4,6,8,12,12a-hexahydro-2H-pyrido[1',2':4,5]pyrazino[2,1-b][1,3]oxazine-9-carboxamide 98%

Supplier: Ambeed
Warning

A374780-100MG A374780-1G A374780-5MG A374780-10MG A374780-2MG A374780-250MG A374780-50MG
76989-912EA 26 USD
76989-912 76989-914 76989-916 77613-128 77613-130 77885-989 77885-990
(4R,12aS)-N-(2,4-Difluorobenzyl)-7-hydroxy-4-methyl-6,8-dioxo-3,4,6,8,12,12a-hexahydro-2H-pyrido[1',2':4,5]pyrazino[2,1-b][1,3]oxazine-9-carboxamide 98%
(4R,12aS)-N-(2,4-Difluorobenzyl)-7-hydroxy-4-methyl-6,8-dioxo-3,4,6,8,12,12a-hexahydro-2H-pyrido[1',2':4,5]pyrazino[2,1-b][1,3]oxazine-9-carboxamide

Precaución: for Research Use Only

Formula: C₂₀H₁₉F₂N₃O₅
MDL Number: MFCD20488027
CAS Number: 1051375-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR