1-Amino-3-chloropropan-2-ol hydrochloride 95%

Supplier: Ambeed
Warning

A374162-250MG A374162-5G
76989-910EA 82.76 USD
76989-910 77594-116
1-Amino-3-chloropropan-2-ol hydrochloride 95%
1-Amino-3-chloropropan-2-ol hydrochloride

Precaución: for Research Use Only

Formula: C₃H₉Cl₂NO
MW: 146.01 g/mol
MDL Number: MFCD01697322
CAS Number: 62037-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR