Benzo[b]thiophene-6-carbonitrile 95%

Supplier: Ambeed
Warning

A373929-100MG A373929-1G A373929-250MG
77033-320EA 35.01 USD
77033-320 77033-322 77033-324
Benzo[b]thiophene-6-carbonitrile 95%
Benzo[b]thiophene-6-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₅NS
MW: 159.208 g/mol
MDL Number: MFCD16877454
CAS Number: 154650-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR