N-Boc-DL-phenylalaninol 97%

Supplier: Ambeed
Warning

A373817-10G A373817-1G A373817-5G
77003-546EA 54 USD
77003-546 77003-548 77003-550 77003-552
N-Boc-DL-phenylalaninol 97%
N-Boc-DL-phenylalaninol

Precaución: for Research Use Only

Formula: C₁₄H₂₁NO₃
MDL Number: MFCD01457313
CAS Number: 145149-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR