2-Methyl-4-(trifluoromethyl)phenylboronic acid 96%

Supplier: Ambeed
Warning

A373468-1G A373468-250MG A373468-5G
77033-314EA 63.66 USD
77033-314 77033-316 77033-318
2-Methyl-4-(trifluoromethyl)phenylboronic acid 96%
2-Methyl-4-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₂
MW: 203.9541 g/mol
MDL Number: MFCD08689550
CAS Number: 957034-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR