2-Chloro-5-fluoro-4-pyridineboronic acid 98%

Supplier: Ambeed
Warning

A373309-1G A373309-250MG A373309-100MG A373309-10G A373309-5G
76989-902EA 20 USD
76989-902 76989-904 77890-763 77890-764 77890-765
2-Chloro-5-fluoro-4-pyridineboronic acid 98%
2-Chloro-5-fluoro-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₄BClFNO₂
MDL Number: MFCD04972404
CAS Number: 951677-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR