5,6,7,8-Tetrahydro-2-naphthol 97%

Supplier: Ambeed
Warning

A373144-10G A373144-25G A373144-5G
77003-534EA 28.65 USD
77003-534 77003-536 77003-538
5,6,7,8-Tetrahydro-2-naphthol 97%
5,6,7,8-Tetrahydro-2-naphthol

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MW: 148.2 g/mol
MDL Number: MFCD00001738
CAS Number: 1125-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR