β-D-Maltose octaacetate 98%

Supplier: Ambeed
Warning

Synonyms: (2S,3R,4S,5R,6R)-6-(Acetoxymethyl)-5-(((2R,3R,4S,5R,6R)-3,4,5-triacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2,3,4-triyl triacetate

A372998-25G A372998-5G A372998-100G
77003-530EA 22 USD
77003-530 77003-532 77882-003
β-D-Maltose octaacetate 98%
beta-D-Maltose octaacetate

Precaución: for Research Use Only

Formula: C₂₈H₃₈O₁₉
MDL Number: MFCD00079432
CAS Number: 22352-19-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR