4-Chloro-3-fluorobenzoyl chloride ≥95%

Supplier: Ambeed
Danger

A372872-1G A372872-5G
77003-526EA 35 USD
77003-526 77003-528
4-Chloro-3-fluorobenzoyl chloride ≥95%
4-Chloro-3-fluorobenzoyl chloride

Precaución: for Research Use Only

Formula: C₇H₃Cl₂FO
MDL Number: MFCD02091532
CAS Number: 160658-68-4
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR