tert-Butyl 6-chloro-2,3-dihydro-1H-pyrrolo[3,2-c]pyridine-1-carboxylate 95%

Supplier: Ambeed
Warning

A372781-100MG A372781-250MG
77016-016EA 151.71 USD
77016-016 77016-018
tert-Butyl 6-chloro-2,3-dihydro-1H-pyrrolo[3,2-c]pyridine-1-carboxylate 95%
tert-Butyl 6-chloro-2,3-dihydro-1H-pyrrolo[3,2-c]pyridine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClN₂O₂
MW: 254.71 g/mol
MDL Number: MFCD27922932
CAS Number: 1649470-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR