6-Chloro-1,2,3,4-tetrahydro-2,7-naphthyridine hydrochloride 96%
Supplier: Ambeed
|
Warning
|
A372390-100MG
A372390-250MG
A372390-50MG
A372390-1G
77016-008EA
223
USD
77016-008
77016-010
77016-012
77890-761
6-Chloro-1,2,3,4-tetrahydro-2,7-naphthyridine hydrochloride 96%
6-Chloro-1,2,3,4-tetrahydro-2,7-naphthyridine hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₀Cl₂N₂ |
MDL Number:
MFCD20528351 CAS Number: 1335053-26-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...