t-Butyl 2-(4-bromophenylsulfonamido)ethylcarbamate 98%

Supplier: Ambeed
Warning

Synonyms: N-(2-BOC-Aminoethyl)-4-bromobenzenesulphonamide, tert-Butyl (2-(4-bromophenylsulphonamido)ethyl)carbamate

A372343-1G A372343-5G
76989-898EA 12.74 USD
76989-898 76989-900
t-Butyl 2-(4-bromophenylsulfonamido)ethylcarbamate 98%
t-Butyl 2-(4-bromophenylsulfonamido)ethylcarbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₉BrN₂O₄S
MW: 379.27 g/mol
MDL Number: MFCD06659915
CAS Number: 310480-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR