Methyl 4-hydroxy-2-methylbenzoate 95%

Supplier: Ambeed
Warning

A371978-100G A371978-10G A371978-1G A371978-250MG A371978-25G A371978-5G
77003-500EA 234 USD
77003-500 77003-502 77003-504 77003-506 77003-508 77003-510
Methyl 4-hydroxy-2-methylbenzoate 95%
Methyl 4-hydroxy-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MDL Number: MFCD09752667
CAS Number: 57556-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR